3-Bromo-5-cyanobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-5-formylbenzonitrile

20416-1G 20416-25G 20416-500MG 20416-5G
76910-156EA 155.96 USD
76910-156 76910-158 76910-160 76910-162
3-Bromo-5-cyanobenzaldehyde ≥97%
3-Bromo-5-cyanobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄BrNO
MW: 210.0275 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09261040
CAS Number: 644982-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR