3-Cyano-5-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

20411-1G 20411-25G 20411-500MG 20411-5G
76910-142EA 201.58 USD
76910-142 76910-144 76910-146 76910-148
3-Cyano-5-methylphenylboronic acid ≥97%
3-Cyano-5-methylphenylboronic acid
Formula: C₈H₈BNO₂
MW: 160.966 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18394418
CAS Number: 1451391-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR