4-Bromo-2-chloro-1-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

20398-25G 20398-5G
76910-134EA 225.98 USD
76910-134 76910-136
4-Bromo-2-chloro-1-isopropoxybenzene ≥97%
4-Bromo-2-chloro-1-isopropoxybenzene
Formula: C₉H₁₀BrClO
Storage Temperature: Ambient
MDL Number: MFCD00070757
CAS Number: 201849-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR