3-Bromo-2,4-dimethylaniline ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-2,4-dimethyl-benzenamine

20334-1G 20334-25G 20334-500MG 20334-5G
76910-110EA 207.94 USD
76910-110 76910-112 76910-114 76910-116
3-Bromo-2,4-dimethylaniline ≥97%
3-Bromo-2,4-dimethylaniline
Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20528102
CAS Number: 66314-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR