4-Bromo-1-fluoro-2-(1-methylethoxy)-benzene ≥97%

Supplier: AOBChem USA

Warning

20323-25G 20323-5G
76910-102EA 339.49 USD
76910-102 76910-104
4-Bromo-1-fluoro-2-(1-methylethoxy)-benzene ≥97%
4-Bromo-1-fluoro-2-(1-methylethoxy)-benzene
Formula: C₉H₁₀BrFO
MW: 233.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18447602
CAS Number: 944278-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR