3-Bromo-5-ethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

20320-1G 20320-25G 20320-5G
76910-090EA 91.24 USD
76910-090 76910-092 76910-094
3-Bromo-5-ethoxybenzoic acid ≥97%
3-Bromo-5-ethoxybenzoic acid
Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17015802
CAS Number: 855198-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR