3-Bromo-5-isopropoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

20317-1G 20317-25G 20317-500MG 20317-5G
76910-078EA 213.25 USD
76910-078 76910-080 76910-082 76910-084
3-Bromo-5-isopropoxybenzaldehyde ≥95%
3-Bromo-5-isopropoxybenzaldehyde
Formula: C₁₀H₁₁BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD17012000
CAS Number: 1112210-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR