4-Bromo-2,5-difluorobenzeneboronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
4-Bromo-2,5-bifluorophenylboronic acid, (4-bromo-2,5-difluorophenyl)boronic acid
20301-25G
20301-5G
76910-050EA
471.04
USD
76910-050
76910-052
4-Bromo-2,5-difluorobenzeneboronic acid ≥97%
4-Bromo-2,5-difluorobenzeneboronic acid
Formula:
C₆H₄BBrF₂O₂ MW: 236.807 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07644467 CAS Number: 1106676-82-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...