3-Methylthioanisole ≥97%

Supplier: AOBChem USA

Warning

20285-25G 20285-5G
76910-044EA 212.18 USD
76910-044 76910-046
3-Methylthioanisole ≥97%
3-Methylthioanisole

Precaución: for Research Use Only

Formula: C₇H₇NO₂S
Storage Temperature: Ambient
MDL Number: MFCD01683019
CAS Number: 4886-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR