4-Ethoxymethylphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

20234-1G 20234-25G 20234-500MG 20234-5G
76910-026EA 126.25 USD
76910-026 76910-028 76910-030 76910-032
4-Ethoxymethylphenylboronic acid ≥95%
4-Ethoxymethylphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD08701751
CAS Number: 279262-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR