4-Bromo-1-fluoro-2-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

20224-25G 20224-5G
76909-998EA 537.88 USD
76909-998 76910-000
4-Bromo-1-fluoro-2-(methoxymethoxy)benzene ≥97%
4-Bromo-1-fluoro-2-(methoxymethoxy)benzene
Formula: C₈H₈BrFO₂
MW: 235.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27756649
CAS Number: 1185836-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR