3-Carboxy-4,5-difluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

20151-1G 20151-25G 20151-500MG 20151-5G
76909-946EA 379.81 USD
76909-946 76909-948 76909-950 76909-952
3-Carboxy-4,5-difluorophenylboronic acid ≥97%
3-Carboxy-4,5-difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BF₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD12546485
CAS Number: 1217500-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR