3-Fluoro-4-iodophenol ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Floro-4-iodophenol

20129-1G 20129-25G 20129-5G
76909-932EA 102.91 USD
76909-932 76909-934 76909-936
3-Fluoro-4-iodophenol ≥97% research grade
3-Fluoro-4-iodophenol
Formula: C₆H₄FIO
Storage Temperature: Refrigerator
MDL Number: MFCD12407087
CAS Number: 122927-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR