3,4-Difluoro-2-ethoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

20060-1G 20060-25G 20060-500MG 20060-5G
76909-916EA 292.81 USD
76909-916 76909-918 76909-920 76909-922
3,4-Difluoro-2-ethoxyphenylboronic acid ≥97%
3,4-Difluoro-2-ethoxyphenylboronic acid
Formula: C₈H₉BF₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD19105367
CAS Number: 1451391-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR