3,5-Dibromo-2-fluorotoluene ≥97%

Supplier: AOBChem USA

Warning

20051-1G 20051-25G 20051-500MG 20051-5G
76909-902EA 236.59 USD
76909-902 76909-904 76909-906 76909-908
3,5-Dibromo-2-fluorotoluene ≥97%
3,5-Dibromo-2-fluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅Br₂F
Storage Temperature: Ambient
MDL Number: MFCD09800763
CAS Number: 85233-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR