3,5-Dichloro-4-propoxyphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

19904-25G 19904-5G
76909-878EA 559.1 USD
76909-878 76909-880
3,5-Dichloro-4-propoxyphenylboronic acid ≥95%
3,5-Dichloro-4-propoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BCl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD15143465
CAS Number: 1218790-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR