3,5-Dichloro-4-hydroxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3,5-Dichloro-4-hydroxyphenyl)boronic acid

19899-1G 19899-25G 19899-500MG 19899-5G
76909-864EA 230.22 USD
76909-864 76909-866 76909-868 76909-870
3,5-Dichloro-4-hydroxyphenylboronic acid ≥97%
3,5-Dichloro-4-hydroxyphenylboronic acid
Formula: C₆H₅BCl₂O₃
MW: 206.819 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19105364
CAS Number: 1335048-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR