3-Bromo-2-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-2-methyl-benzenemethanol, (3-Bromo-o-tolyl)methanol, (3-Bromo-2-methylphenyl)methanol

19817-25G 19817-5G
76909-832EA 247.19 USD
76909-832 76909-834
3-Bromo-2-methylbenzyl alcohol ≥97%
3-Bromo-2-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉BrO
MW: 201.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847531
CAS Number: 83647-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR