4-Bromo-2-chloro-N,N-dimethylaniline ≥97%

Supplier: AOBChem USA

Warning

19778-1G 19778-25G 19778-500MG 19778-5G
76909-816EA 215.37 USD
76909-816 76909-818 76909-820 76909-822
4-Bromo-2-chloro-N,N-dimethylaniline ≥97%
4-Bromo-2-chloro-N,N-dimethylaniline
Formula: C₈H₉BrClN
Storage Temperature: Refrigerator
MDL Number: MFCD07780653
CAS Number: 50638-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR