4-(2-Pyridylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(Pyridin-2-yl)aminocarbonylphenylboronic acid, (4-(Pyridin-2-ylcarbamoyl)phenyl)boronic acid, 4-(2-Pyridylcarbamoyl)benzeneboronic acid

19758-1G 19758-25G 19758-500MG 19758-5G
76909-800EA 177.18 USD
76909-800 76909-802 76909-804 76909-806
4-(2-Pyridylcarbamoyl)phenylboronic acid ≥97%
4-(2-Pyridylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₁BN₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD06659876
CAS Number: 850568-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR