3-Carboxy-2,4-difluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

19581-1G 19581-25G 19581-5G
76909-774EA 62.6 USD
76909-774 76909-776 76909-778
3-Carboxy-2,4-difluorophenylboronic acid ≥97%
3-Carboxy-2,4-difluorophenylboronic acid
Formula: C₇H₅BF₂O₄
Storage Temperature: Ambient
MDL Number: MFCD16295273
CAS Number: 1451393-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR