3,5-Dibromo-4-iodopyridine ≥95%

Supplier: AOBChem USA

Warning

19561-1G 19561-25G 19561-5G
76909-760EA 62.6 USD
76909-760 76909-762 76909-764
3,5-Dibromo-4-iodopyridine ≥95%
3,5-Dibromo-4-iodopyridine
Formula: C₅H₂Br₂IN
MW: 362.788 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185522
CAS Number: 1214383-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR