3,5-Dichloro-4-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

19466-1G 19466-25G 19466-500MG 19466-5G
76909-706EA 245.07 USD
76909-706 76909-708 76909-710 76909-712
3,5-Dichloro-4-formylphenylboronic acid ≥97%
3,5-Dichloro-4-formylphenylboronic acid
Formula: C₇H₅BCl₂O₃
MW: 218.83 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16295268
CAS Number: 1451393-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR