(3-Bromo-5-isopropylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

19422-1G 19422-25G 19422-5G
76909-684EA 94.43 USD
76909-684 76909-686 76909-688
(3-Bromo-5-isopropylphenyl)boronic acid ≥97%
(3-Bromo-5-isopropylphenyl)boronic acid
Formula: C₉H₁₂BBrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD16295265
CAS Number: 1451390-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR