4-(Benzyloxy)-3,5-difluorophenol ≥97%

Supplier: AOBChem USA

Warning

19340-1G 19340-25G 19340-500MG 19340-5G
76909-668EA 266.29 USD
76909-668 76909-670 76909-672 76909-674
4-(Benzyloxy)-3,5-difluorophenol ≥97%
4-(Benzyloxy)-3,5-difluorophenol
Formula: C₁₃H₁₀F₂O₂
MW: 236.214 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29044713
CAS Number: 99045-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR