3,6-Dimethoxypyridazine-4-boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3,6-Dimethoxylpyridazine-4-boronic acid

19208-1G 19208-25G 19208-5G
76909-614EA 64.72 USD
76909-614 76909-616 76909-618
3,6-Dimethoxypyridazine-4-boronic acid ≥97%
3,6-Dimethoxypyridazine-4-boronic acid

Precaución: for Research Use Only

Formula: C₆H₉BN₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD10700157
CAS Number: 1015480-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR