(3-Chloro-2-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-2-methylbenzyl alcohol

19157-25G 19157-5G
76909-608EA 601.54 USD
76909-608 76909-610
(3-Chloro-2-methylphenyl)methanol ≥97%
(3-Chloro-2-methylphenyl)methanol
Formula: C₈H₉ClO
MW: 156.61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09753680
CAS Number: 90369-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR