2-Bromo-1-chloro-4-isopropylbenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-4-chloroisopropylbenzene

19120-1G 19120-25G 19120-5G
76909-578EA 81.69 USD
76909-578 76909-580 76909-582
2-Bromo-1-chloro-4-isopropylbenzene ≥97%
2-Bromo-1-chloro-4-isopropylbenzene
Formula: C₉H₁₀BrCl
MW: 233.53 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11845950
CAS Number: 90350-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR