3-Bromo-2-fluoro-4-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

19108-1G 19108-25G 19108-500MG 19108-5G
76909-562EA 409.51 USD
76909-562 76909-564 76909-566 76909-568
3-Bromo-2-fluoro-4-methoxybenzaldehyde ≥97%
3-Bromo-2-fluoro-4-methoxybenzaldehyde
Formula: C₈H₆BrFO₂
MW: 233.034 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27959302
CAS Number: 1155877-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR