3-Bromo-2-fluoro-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

19065-25G 19065-5G
76909-554EA 516.66 USD
76909-554 76909-556
3-Bromo-2-fluoro-4-methylpyridine ≥97%
3-Bromo-2-fluoro-4-methylpyridine
Formula: C6H5BrFN
Storage Temperature: Refrigerator
MDL Number: MFCD08277289
CAS Number: 884495-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR