3,5-Dibromothiophene-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

18907-25G 18907-5G
76909-522EA 453.01 USD
76909-522 76909-524
3,5-Dibromothiophene-2-carboxylic acid ≥97%
3,5-Dibromothiophene-2-carboxylic acid
Formula: C₅H₂Br₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD01006744
CAS Number: 7311-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR