3-Aminocarbonyl-2-fluorophenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

18882-1G 18882-25G 18882-500MG 18882-5G
76909-504EA 350.1 USD
76909-504 76909-506 76909-508 76909-510
3-Aminocarbonyl-2-fluorophenylboronic acid ≥97% research grade
3-Aminocarbonyl-2-fluorophenylboronic acid
Formula: C₇H₇BFNO₃
Storage Temperature: Ambient
MDL Number: MFCD16295255
CAS Number: 1451392-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR