5-Methylpyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Hydroxy-5-methylpyridine, 3-Hydroxy-5-picoline

18709-25G 18709-5G
76909-462EA 307.67 USD
76909-462 76909-464
5-Methylpyridin-3-ol ≥97%
5-Methylpyridin-3-ol

Precaución: for Research Use Only

Formula: C₆H₇NO
MW: 109.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00661297
CAS Number: 42732-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR