5-Bromo-6-methylpyridine-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-6-methylpicolinic acid, 3-Bromo-2-methylpyridine-6-carboxylic acid

18637-1G 18637-25G 18637-5G
76909-442EA 84.88 USD
76909-442 76909-444 76909-446
5-Bromo-6-methylpyridine-2-carboxylic acid ≥97%
5-Bromo-6-methylpyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MW: 216.032 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13181588
CAS Number: 137778-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR