(3-Bromo-2-methylphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-2-methylphenylboronic acid

18609-1G 18609-25G 18609-5G
76909-430EA 112.46 USD
76909-430 76909-432 76909-434
(3-Bromo-2-methylphenyl)boronic acid ≥97% research grade
(3-Bromo-2-methylphenyl)boronic acid
Formula: C₇H₈BBrO₂
MW: 214.85 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18447597
CAS Number: 1184298-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR