3-Bromo-6-fluoro-2-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

18601-1G 18601-25G 18601-500MG 18601-5G
76909-404EA 200.52 USD
76909-404 76909-406 76909-408 76909-410
3-Bromo-6-fluoro-2-hydroxybenzaldehyde ≥97%
3-Bromo-6-fluoro-2-hydroxybenzaldehyde
Formula: C₇H₄BrFO₂
MW: 219.0079 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20528359
CAS Number: 199287-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR