3-Chloro-2-fluoro-6-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

18595-1G 18595-500MG 18595-5G
76909-392EA 755.37 USD
76909-392 76909-394 76909-396
3-Chloro-2-fluoro-6-hydroxybenzaldehyde ≥97%
3-Chloro-2-fluoro-6-hydroxybenzaldehyde
Formula: C₇H₄ClFO₂
MW: 174.557 g/mol
Storage Temperature: Refrigerator
CAS Number: 775334-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR