3-Bromo-2-fluoro-6-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

18591-1G 18591-25G 18591-500MG 18591-5G
76909-376EA 290.69 USD
76909-376 76909-378 76909-380 76909-382
3-Bromo-2-fluoro-6-methoxybenzaldehyde ≥97%
3-Bromo-2-fluoro-6-methoxybenzaldehyde
Formula: C₈H₆BrFO₂
MW: 233.03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15528066
CAS Number: 1160653-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR