(3-Chloro-4-(methylthio)phenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-4-(methylthio)phenylboronic acid

18505-1G 18505-25G 18505-5G
76909-344EA 114.58 USD
76909-344 76909-346 76909-348
(3-Chloro-4-(methylthio)phenyl)boronic acid ≥97% research grade
(3-Chloro-4-(methylthio)phenyl)boronic acid
Formula: C₇H₈BClO₂S
Storage Temperature: Ambient
MDL Number: MFCD22543649
CAS Number: 877383-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR