3-Chloro-4-iodoanisole ≥97%

Supplier: AOBChem USA

Warning

18487-1G 18487-25G 18487-500MG 18487-5G
76909-330EA 393.6 USD
76909-330 76909-332 76909-334 76909-336
3-Chloro-4-iodoanisole ≥97%
3-Chloro-4-iodoanisole
Formula: C₇H₆ClIO
MW: 268.479 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18393317
CAS Number: 219735-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR