3-Carboxy-2-chloropyridine-5-boronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

18445-1G 18445-25G 18445-500MG 18445-5G
76909-314EA 379.81 USD
76909-314 76909-316 76909-318 76909-320
3-Carboxy-2-chloropyridine-5-boronic acid ≥95% research grade
3-Carboxy-2-chloropyridine-5-boronic acid
Formula: C₆H₅BClNO₄
MW: 201.372 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13181645
CAS Number: 1451393-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR