[4-(methoxycarbonyl)-3-methylphenyl]boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-(Methoxycarbonyl)-3-methylphenyl)boronic acid, 3-Methyl-4-(methoxycarbonyl)phenylboronic acid

18430-1G 18430-25G 18430-500MG 18430-5G
76909-298EA 138.98 USD
76909-298 76909-300 76909-302 76909-304
[4-(methoxycarbonyl)-3-methylphenyl]boronic acid ≥97%
[4-(methoxycarbonyl)-3-methylphenyl]boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₄
MW: 193.992 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18432535
CAS Number: 603122-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR