3,5-Dichloro-4-ethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

18417-1G 18417-25G 18417-500MG 18417-5G
76909-288EA 282.2 USD
76909-288 76909-290 76909-292 76909-294
3,5-Dichloro-4-ethoxybenzaldehyde ≥97%
3,5-Dichloro-4-ethoxybenzaldehyde
Formula: C₉H₈Cl₂O₂
MW: 219.065 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02611618
CAS Number: 43171-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR