3,5-Dibromo-4-ethylpyridine ≥97% research grade

Supplier: AOBChem USA

Warning

18411-1G 18411-25G 18411-5G
76909-278EA 103.97 USD
76909-278 76909-280 76909-282
3,5-Dibromo-4-ethylpyridine ≥97% research grade
3,5-Dibromo-4-ethylpyridine
Formula: C₇H₇Br₂N
MW: 264.945 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19440791
CAS Number: 125419-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR