4-Bromo-2,6-dimethylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Bromo-2,6-dimethylphenyl)boronic acid

18384-1G 18384-25G 18384-5G
76909-268EA 117.76 USD
76909-268 76909-270 76909-272
4-Bromo-2,6-dimethylphenylboronic acid ≥97%
4-Bromo-2,6-dimethylphenylboronic acid
Formula: C₈H₁₀BBrO₂
MW: 228.879 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13181651
CAS Number: 1160561-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR