3-Chloro-5-fluoro-4-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

18371-25G 18371-5G
76909-258EA 275.84 USD
76909-258 76909-260
3-Chloro-5-fluoro-4-methoxybenzaldehyde ≥97%
3-Chloro-5-fluoro-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
MW: 188.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115863
CAS Number: 177994-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR