3,5-Dibromo-1-(trimethylsilyl)benzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3,5-Dibromophenyltrimethylsilane, (3,5-Dibromophenyl)trimethylsilane

18275-25G 18275-5G
76909-234EA 403.15 USD
76909-234 76909-236
3,5-Dibromo-1-(trimethylsilyl)benzene ≥97%
3,5-Dibromo-1-(trimethylsilyl)benzene

Precaución: for Research Use Only

Formula: C₉H₁₂Br₂Si
MW: 308.0851 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03844787
CAS Number: 17878-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR