3-Bromo-5-fluoro-pyridine-4-carboxylic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-5-fluoroisonicotinic acid, 3-Bromo-5-fluoropyridine-4-carboxylic acid

18216-1G 18216-25G 18216-5G
76909-222EA 97.61 USD
76909-222 76909-224 76909-226
3-Bromo-5-fluoro-pyridine-4-carboxylic acid ≥97% research grade
3-Bromo-5-fluoro-pyridine-4-carboxylic acid
Formula: C₆H₃BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD13181633
CAS Number: 955372-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR