3,5-Bis(trifluoromethyl)-2-bromophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

18170-1G 18170-25G 18170-500MG 18170-5G
76909-192EA 182.48 USD
76909-192 76909-194 76909-196 76909-198
3,5-Bis(trifluoromethyl)-2-bromophenylboronic acid ≥97%
3,5-Bis(trifluoromethyl)-2-bromophenylboronic acid
Formula: C₈H₄BBrF₆O₂
MW: 336.822 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16295250
CAS Number: 1451393-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR