3-Bromo-5-ethylpyridine ≥97% research grade

Supplier: AOBChem USA

Warning

18101-1G 18101-25G 18101-5G
76909-178EA 73.21 USD
76909-178 76909-180 76909-182
3-Bromo-5-ethylpyridine ≥97% research grade
3-Bromo-5-ethylpyridine
Formula: C7H8BrN
Storage Temperature: Ambient
MDL Number: MFCD09800592
CAS Number: 142337-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR